Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015002.2 | NP_055817.1 | 4 - 5 | 632 | 69 |
NM_033624.2 | NP_296373.1 | 4 - 5 | 632 | 69 | |
XM_017019037.1 | XP_016874526.1 | 4 - 5 | 637 | 69 | |
XM_017019038.1 | XP_016874527.1 | 4 - 5 | 637 | 69 | |
XM_017019039.1 | XP_016874528.1 | 3 - 4 | 248 | 69 | |
GenBank mRNA | AB020682.1 | - | 4 - 5 | 594 | 69 |
AF174601.1 | - | 4 - 5 | 593 | 69 | |
AK001699.1 | - | 4 - 5 | 616 | 69 | |
AK074714.1 | - | 1 - 2 | 101 | 69 | |
AK314573.1 | - | 4 - 5 | 605 | 69 | |
AL136899.1 | - | 4 - 5 | 445 | 69 | |
BC034045.1 | - | 5 - 6 | 559 | 69 | |
BC042333.1 | - | 5 - 6 | 532 | 69 | |
BC091496.1 | - | 4 - 5 | 591 | 69 |