Assay Design



Gene Transcripts

Gene Symbol
FBXO21
Entrez Gene ID
23014
Gene Name
F-box protein 21
Gene Aliases
FBX21
Chromosome Location
Chr.12: 117143473 - 117190500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015002.2 NP_055817.1 9 - 10 1369 97
XM_017019038.1 XP_016874527.1 8 - 9 1194 97
XM_017019039.1 XP_016874528.1 8 - 9 985 97
GenBank mRNA AB020682.1 - 9 - 10 1331 97
AF174601.1 - 9 - 10 1330 97
AK001699.1 - 9 - 10 1353 97
AK074714.1 - 6 - 7 838 97
AK308141.1 - 10 - 11 1275 97
AK314573.1 - 9 - 10 1342 97
BC034045.1 - 10 - 11 1296 97
BC042333.1 - 9 - 10 1089 97
BC091496.1 - 9 - 10 1328 97

View Full Product Details