Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111322.1 | NP_001104792.1 | 8 - 9 | 906 | 67 |
NM_024072.3 | NP_076977.3 | 8 - 9 | 906 | 67 | |
GenBank mRNA | AF478457.1 | - | 8 - 9 | 906 | 67 |
AY148094.1 | - | 8 - 9 | 885 | 67 | |
BC005848.2 | - | 8 - 9 | 827 | 67 |