Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032279.1 | NP_001027450.1 | 3 - 4 | 410 | 75 |
NM_005133.2 | NP_005124.1 | 3 - 4 | 414 | 75 | |
GenBank mRNA | AF121951.1 | - | 3 - 4 | 414 | 75 |
AL713740.1 | - | 2 - 3 | 1109 | 75 | |
BC052622.1 | - | 3 - 4 | 371 | 75 | |
BC093726.1 | - | 3 - 4 | 410 | 75 | |
BC093728.1 | - | 3 - 4 | 409 | 75 | |
Y13835.2 | - | 3 - 4 | 383 | 75 |