Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142536.1 | NP_001136008.1 | 5 - 6 | 944 | 101 |
NM_001142538.1 | NP_001136010.1 | 4 - 5 | 1034 | 101 | |
GenBank mRNA | AF244976.1 | - | 5 - 6 | 913 | 101 |
AF244977.1 | - | 4 - 5 | 1034 | 101 |