Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003286.2 | NP_003277.1 | 9 - 10 | 978 | 90 |
GenBank mRNA | AK292943.1 | - | 9 - 10 | 977 | 90 |
BC136297.1 | - | 9 - 10 | 829 | 90 | |
J03250.1 | - | 9 - 10 | 943 | 90 | |
U07804.1 | - | 9 - 10 | 722 | 90 | |
U07806.1 | - | 9 - 10 | 722 | 90 |