Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080675.3 | NP_542406.2 | 10 - 11 | 821 | 102 |
XM_011528573.1 | XP_011526875.1 | 11 - 12 | 905 | 102 | |
XM_011528574.1 | XP_011526876.1 | 9 - 10 | 761 | 102 | |
XM_011528575.1 | XP_011526877.1 | 7 - 8 | 540 | 102 | |
GenBank mRNA | AF401350.1 | - | 10 - 11 | 821 | 102 |
BC026118.2 | - | 10 - 11 | 821 | 102 | |
BC029528.1 | - | 10 - 11 | 814 | 102 |