Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159920.1 | NP_001153392.1 | 11 - 12 | 1840 | 72 |
NM_001160030.1 | NP_001153502.1 | 11 - 12 | 1840 | 72 | |
NM_002019.4 | NP_002010.2 | 11 - 12 | 1840 | 72 | |
XM_011535014.1 | XP_011533316.1 | 11 - 12 | 1840 | 72 | |
XM_017020485.1 | XP_016875974.1 | 11 - 12 | 1840 | 72 | |
GenBank mRNA | AF063657.2 | - | 11 - 12 | 1555 | 72 |
AK292936.1 | - | 11 - 12 | 1840 | 72 | |
AK300392.1 | - | 11 - 12 | 1840 | 72 | |
AK309901.1 | - | 11 - 12 | 1840 | 72 | |
BC039007.1 | - | 11 - 12 | 1791 | 72 | |
EU360600.1 | - | 11 - 12 | 1840 | 72 | |
EU368830.1 | - | 11 - 12 | 1555 | 72 | |
S77812.1 | - | 1 - 2 | 49 | 72 | |
U01134.1 | - | 11 - 12 | 1804 | 72 |