Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002019.4 | NP_002010.2 | 28 - 29 | 4006 | 69 |
XM_017020485.1 | XP_016875974.1 | 27 - 28 | 3892 | 69 | |
GenBank mRNA | AB209050.1 | - | 12 - 13 | 1362 | 69 |
AF063657.2 | - | 28 - 29 | 3721 | 69 | |
BU151621.1 | - | 5 - 6 | 544 | 69 | |
DQ836394.1 | - | 14 - 15 | 1546 | 69 | |
DQ836395.1 | - | 15 - 16 | 1608 | 69 | |
EF491868.1 | - | 10 - 11 | 1255 | 69 | |
EF491869.1 | - | 7 - 8 | 1042 | 69 | |
EF491870.1 | - | 8 - 9 | 946 | 69 | |
JF509744.1 | - | 10 - 11 | 1139 | 69 |