Assay Design



Gene Transcripts

Gene Symbol
SLK
Entrez Gene ID
9748
Gene Name
STE20 like kinase
Gene Aliases
LOSK, STK2, bA16H23.1, se20-9
Chromosome Location
Chr.10: 103967173 - 104029233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014720.3 NP_055535.2 12 - 13 3338 98
XM_011540401.2 XP_011538703.1 11 - 12 1994 98
GenBank mRNA AF273048.1 - 12 - 13 2811 98

View Full Product Details