Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323977.1 | NP_001310906.1 | 4 - 5 | 412 | 86 |
NM_001323978.1 | NP_001310907.1 | 5 - 6 | 840 | 86 | |
NM_001323979.1 | NP_001310908.1 | 4 - 5 | 527 | 86 | |
NM_001323980.1 | NP_001310909.1 | 4 - 5 | 757 | 86 | |
NM_001323981.1 | NP_001310910.1 | 5 - 6 | 970 | 86 | |
NM_194298.2 | NP_919274.1 | 4 - 5 | 887 | 86 | |
XM_017015883.1 | XP_016871372.1 | 4 - 5 | 887 | 86 | |
XM_017015884.1 | XP_016871373.1 | 5 - 6 | 1220 | 86 | |
GenBank mRNA | AK125791.1 | - | 4 - 5 | 547 | 86 |
BC126238.1 | - | 3 - 4 | 476 | 86 | |
BC130567.1 | - | 3 - 4 | 476 | 86 | |
DA759951.1 | - | 4 - 5 | 412 | 86 |