Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256910.1 | NP_001243839.1 | 3 - 4 | 830 | 114 |
NM_004728.3 | NP_004719.2 | 3 - 4 | 711 | 114 | |
XM_011540336.1 | XP_011538638.1 | 3 - 4 | 703 | 114 | |
XM_017016910.1 | XP_016872399.1 | 3 - 4 | 621 | 114 | |
GenBank mRNA | AK315585.1 | - | 3 - 4 | 660 | 114 |
BC008071.2 | - | 3 - 4 | 649 | 114 | |
BX648405.1 | - | 3 - 4 | 114 | ||
CR749598.1 | - | 3 - 4 | 831 | 114 | |
U41387.1 | - | 3 - 4 | 662 | 114 |