Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256910.1 | NP_001243839.1 | 6 - 7 | 1312 | 101 |
NM_004728.3 | NP_004719.2 | 6 - 7 | 1193 | 101 | |
XM_011540336.1 | XP_011538638.1 | 6 - 7 | 1185 | 101 | |
XM_017016910.1 | XP_016872399.1 | 6 - 7 | 1103 | 101 | |
GenBank mRNA | AK025629.1 | - | 3 - 4 | 495 | 101 |
AK315585.1 | - | 6 - 7 | 1142 | 101 | |
BC008071.2 | - | 6 - 7 | 1131 | 101 | |
BX648405.1 | - | 6 - 7 | 1079 | 101 | |
CR749598.1 | - | 6 - 7 | 1313 | 101 | |
U41387.1 | - | 6 - 7 | 1144 | 101 |