Assay Design



Gene Transcripts

Gene Symbol
AK7
Entrez Gene ID
122481
Gene Name
adenylate kinase 7
Gene Aliases
AK 7
Chromosome Location
Chr.14: 96392111 - 96489427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152327.3 NP_689540.2 8 - 9 911 79
XM_006720021.2 XP_006720084.1 8 - 9 911 79
XM_011536403.2 XP_011534705.1 8 - 9 911 79
XM_017020956.1 XP_016876445.1 8 - 9 911 79
XM_017020958.1 XP_016876447.1 8 - 9 911 79
XM_017020959.1 XP_016876448.1 8 - 9 911 79
GenBank mRNA AK057426.1 - 8 - 9 911 79
BC035256.1 - 8 - 9 900 79

View Full Product Details