Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004370.5 | NP_004361.3 | 26 - 27 | 4994 | 98 |
NM_080645.2 | NP_542376.2 | 11 - 12 | 1502 | 98 | |
XM_011535434.1 | XP_011533736.1 | 26 - 27 | 5138 | 98 | |
XM_011535435.1 | XP_011533737.1 | 25 - 26 | 4865 | 98 | |
XM_011535436.2 | XP_011533738.1 | 11 - 12 | 1857 | 98 | |
XM_017010252.1 | XP_016865741.1 | 25 - 26 | 5390 | 98 | |
GenBank mRNA | BC146868.1 | - | 11 - 12 | 1446 | 98 |
U73778.1 | - | 26 - 27 | 4941 | 98 |