Assay Design



Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 51 - 52 8113 153
NM_080645.2 NP_542376.2 36 - 37 4621 153
XM_011535434.1 XP_011533736.1 51 - 52 8257 153
XM_011535435.1 XP_011533737.1 50 - 51 7984 153
XM_011535436.2 XP_011533738.1 36 - 37 4976 153
XM_017010252.1 XP_016865741.1 50 - 51 8509 153
GenBank mRNA BC146868.1 - 36 - 37 4565 153
U73778.1 - 51 - 52 8060 153

View Full Product Details