Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178498.3 | NP_848593.2 | 11 - 12 | 1627 | 104 |
XM_006718155.3 | XP_006718218.1 | 11 - 12 | 1530 | 104 | |
XM_011519920.2 | XP_011518222.1 | 12 - 13 | 1107 | 104 | |
XM_017017244.1 | XP_016872733.1 | 12 - 13 | 1553 | 104 | |
GenBank mRNA | AY299482.1 | - | 11 - 12 | 1336 | 104 |
BC041454.1 | - | 11 - 12 | 1100 | 104 |