Assay Design



Gene Transcripts

Gene Symbol
BAAT
Entrez Gene ID
570
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases
BACAT, BAT
Chromosome Location
Chr.9: 101360417 - 101385005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127610.1 NP_001121082.1 1 - 2 47 82
GenBank mRNA AK315347.1 - 1 - 2 92 82

View Full Product Details