Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323505.1 | NP_001310434.1 | 17 - 18 | 2654 | 113 |
NM_001323506.1 | NP_001310435.1 | 17 - 18 | 2657 | 113 | |
NM_003999.2 | NP_003990.1 | 17 - 18 | 2774 | 113 | |
XM_005248384.1 | XP_005248441.1 | 17 - 18 | 2764 | 113 | |
XM_017010020.1 | XP_016865509.1 | 11 - 12 | 1464 | 113 | |
GenBank mRNA | AK304016.1 | - | 11 - 12 | 1700 | 113 |
BC125209.1 | - | 17 - 18 | 2544 | 113 | |
BC125210.1 | - | 17 - 18 | 2544 | 113 | |
U60805.1 | - | 17 - 18 | 2739 | 113 |