Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323505.1 | NP_001310434.1 | 8 - 9 | 1413 | 96 |
NM_001323507.1 | NP_001310436.1 | 8 - 9 | 1413 | 96 | |
NM_003999.2 | NP_003990.1 | 8 - 9 | 1533 | 96 | |
XM_005248387.2 | XP_005248444.1 | 8 - 9 | 1520 | 96 | |
XM_017010019.1 | XP_016865508.1 | 8 - 9 | 1520 | 96 | |
GenBank mRNA | BC125209.1 | - | 8 - 9 | 1303 | 96 |
BC125210.1 | - | 8 - 9 | 1303 | 96 | |
U60805.1 | - | 8 - 9 | 1498 | 96 |