Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014729.2 | NP_055544.1 | 1 - 2 | 328 | 65 |
GenBank mRNA | AB018351.1 | - | 1 - 2 | 330 | 65 |
AK000971.1 | - | 1 - 2 | 110 | 65 | |
AK302331.1 | - | 1 - 2 | 252 | 65 | |
BC016665.1 | - | 1 - 2 | 171 | 65 |