Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004855.4 | NP_004846.4 | 11 - 12 | 102 | |
XM_005254795.4 | XP_005254852.1 | 9 - 10 | 102 | ||
XM_011522235.2 | XP_011520537.1 | 11 - 12 | 102 | ||
XM_011522236.2 | XP_011520538.1 | 10 - 11 | 102 | ||
XM_011522237.2 | XP_011520539.1 | 10 - 11 | 102 | ||
XM_017022730.1 | XP_016878219.1 | 10 - 11 | 102 | ||
XM_017022731.1 | XP_016878220.1 | 11 - 12 | 102 | ||
XM_017022732.1 | XP_016878221.1 | 9 - 10 | 102 | ||
GenBank mRNA | AK093138.1 | - | 2 - 3 | 102 | |
AK223330.1 | - | 11 - 12 | 102 | ||
AK293666.1 | - | 8 - 9 | 102 | ||
AK314706.1 | - | 11 - 12 | 102 | ||
BC017711.1 | - | 11 - 12 | 1771 | 102 | |
D42138.1 | - | 11 - 12 | 102 |