Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002309.4 | NP_002300.1 | 2 - 3 | 356 | 77 |
GenBank mRNA | AK315310.1 | - | 2 - 3 | 304 | 77 |
BC069540.1 | - | 2 - 3 | 250 | 77 | |
BC093733.1 | - | 2 - 3 | 248 | 77 | |
BC093735.1 | - | 2 - 3 | 248 | 77 | |
CN409458.1 | - | 2 - 3 | 356 | 77 | |
X13967.1 | - | 2 - 3 | 244 | 77 |