Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261824.1 | NP_001248753.1 | 5 - 6 | 769 | 150 |
NM_001320795.1 | NP_001307724.1 | 5 - 6 | 972 | 150 | |
NM_003720.3 | NP_003711.1 | 5 - 6 | 784 | 150 | |
NM_203433.2 | NP_982257.1 | 4 - 5 | 721 | 150 | |
NR_049728.1 | 4 - 5 | 677 | 150 | ||
XM_017028488.1 | XP_016883977.1 | 4 - 5 | 635 | 150 | |
GenBank mRNA | AB451468.1 | - | 4 - 5 | 592 | 150 |
AF417108.1 | - | 4 - 5 | 548 | 150 | |
AJ006291.1 | - | 5 - 6 | 718 | 150 | |
AK001455.1 | - | 5 - 6 | 746 | 150 | |
AK308614.1 | - | 4 - 5 | 655 | 150 | |
AK315406.1 | - | 5 - 6 | 686 | 150 | |
AW594615.1 | - | 2 - 3 | 151 | ||
AY463963.1 | - | 4 - 5 | 592 | 150 | |
BC003619.2 | - | 5 - 6 | 701 | 150 | |
BC010424.1 | - | 5 - 6 | 693 | 150 | |
BC011755.2 | - | 5 - 6 | 701 | 150 | |
BC012809.2 | - | 5 - 6 | 701 | 150 | |
BM013494.1 | - | 5 - 6 | 654 | 153 | |
BQ008489.1 | - | 2 - 3 | 150 | ||
CR541821.1 | - | 5 - 6 | 655 | 150 | |
CR541852.1 | - | 5 - 6 | 655 | 150 |