Assay Design



Gene Transcripts

Gene Symbol
CASR
Entrez Gene ID
846
Gene Name
calcium sensing receptor
Gene Aliases
CAR, EIG8, FHH, FIH, GPRC2A, HHC, HHC1, HYPOC1, NSHPT, PCAR1
Chromosome Location
Chr.3: 122183683 - 122286503 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000388.3 NP_000379.2 5 - 6 1981 57
XM_005247837.2 XP_005247894.1 3 - 4 1211 57
XM_006713789.3 XP_006713852.1 5 - 6 2562 57
XM_017007324.1 XP_016862813.1 6 - 7 3927 57
XM_017007325.1 XP_016862814.1 5 - 6 1953 57
GenBank mRNA BC104999.1 - 4 - 5 1633 57
BC112236.1 - 4 - 5 1633 57
D50855.1 - 4 - 5 1637 57
FJ360896.1 - 1 - 2 198 57
S83176.1 - 4 - 5 1609 57
U20759.1 - 5 - 6 1981 57

View Full Product Details