Assay Design



Gene Transcripts

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
Chromosome Location
Chr.7: 38383694 - 38631567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001635.3 NP_001626.1 16 - 17 1489 60
XM_011515271.2 XP_011513573.1 16 - 17 1934 60
XM_017011995.1 XP_016867484.1 16 - 17 3152 60
GenBank mRNA AK291447.1 - 16 - 17 1293 60
BC034376.1 - 16 - 17 1280 60
U07616.1 - 16 - 17 1383 60
X81438.1 - 16 - 17 1347 60

View Full Product Details