Assay Design



Gene Transcripts

Gene Symbol
EEPD1
Entrez Gene ID
80820
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
HSPC107
Chromosome Location
Chr.7: 36153149 - 36301543 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030636.2 NP_085139.2 3 - 4 1574 71
GenBank mRNA AB051493.1 - 3 - 4 1315 71
AK027386.1 - 3 - 4 1546 71
BC065518.1 - 4 - 5 1474 71

View Full Product Details