Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015884.3 | NP_056968.1 | 7 - 8 | 1165 | 125 |
GenBank mRNA | AF019612.1 | - | 7 - 8 | 1067 | 125 |
AK292933.1 | - | 7 - 8 | 1072 | 125 | |
AK312474.1 | - | 7 - 8 | 998 | 125 |