Assay Design



Gene Transcripts

Gene Symbol
ZC3H8
Entrez Gene ID
84524
Gene Name
zinc finger CCCH-type containing 8
Gene Aliases
Fliz1, ZC3HDC8
Chromosome Location
Chr.2: 112215035 - 112255087 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032494.2 NP_115883.2 2 - 3 266 162
XM_017005107.1 XP_016860596.1 2 - 3 219 162
GenBank mRNA AF334161.1 - 2 - 3 252 162
AK315329.1 - 2 - 3 220 162
BC032001.1 - 2 - 3 204 162
BG773518.1 - 2 - 3 295 162

View Full Product Details