Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032494.2 | NP_115883.2 | 6 - 7 | 834 | 119 |
XM_011511998.1 | XP_011510300.1 | 6 - 7 | 779 | 119 | |
XM_017005107.1 | XP_016860596.1 | 6 - 7 | 787 | 119 | |
GenBank mRNA | AK315329.1 | - | 6 - 7 | 787 | 119 |
BC032001.1 | - | 6 - 7 | 772 | 119 | |
CV574571.1 | - | 4 - 5 | 380 | 119 |