Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206855.1 | NP_001193784.1 | 11 - 12 | 1302 | 95 |
NM_003227.3 | NP_003218.2 | 14 - 15 | 1722 | 95 | |
XM_005250553.4 | XP_005250610.1 | 15 - 16 | 2028 | 95 | |
XM_017012573.1 | XP_016868062.1 | 15 - 16 | 1830 | 95 | |
GenBank mRNA | AF067864.1 | - | 14 - 15 | 1708 | 95 |
AK000421.1 | - | 10 - 11 | 1034 | 95 | |
BC142630.1 | - | 14 - 15 | 1708 | 95 | |
BC150192.1 | - | 11 - 12 | 1516 | 95 |