Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206855.1 | NP_001193784.1 | 13 - 14 | 1621 | 77 |
NM_003227.3 | NP_003218.2 | 16 - 17 | 2041 | 77 | |
XM_005250553.4 | XP_005250610.1 | 17 - 18 | 2347 | 77 | |
XM_017012573.1 | XP_016868062.1 | 17 - 18 | 2149 | 77 | |
GenBank mRNA | AF067864.1 | - | 16 - 17 | 2027 | 77 |
AK000421.1 | - | 12 - 13 | 1353 | 77 | |
BC142630.1 | - | 16 - 17 | 2027 | 77 | |
BC150192.1 | - | 12 - 13 | 1920 | 77 |