Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003227.3 | NP_003218.2 | 2 - 3 | 323 | 59 |
XM_005250553.4 | XP_005250610.1 | 3 - 4 | 629 | 59 | |
XM_017012573.1 | XP_016868062.1 | 3 - 4 | 431 | 59 | |
GenBank mRNA | AF067864.1 | - | 2 - 3 | 309 | 59 |
AK022002.1 | - | 3 - 4 | 440 | 59 | |
BC142630.1 | - | 2 - 3 | 309 | 59 | |
BC150192.1 | - | 2 - 3 | 287 | 59 | |
DR004213.1 | - | 2 - 3 | 466 | 59 |