Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014920.3 | NP_055735.1 | 10 - 11 | 1718 | 116 |
NM_016513.4 | NP_057597.2 | 11 - 12 | 1829 | 116 | |
XM_011514419.1 | XP_011512721.1 | 10 - 11 | 1739 | 116 | |
XM_011514420.2 | XP_011512722.1 | 11 - 12 | 1881 | 116 | |
XM_011514421.1 | XP_011512723.1 | 11 - 12 | 1820 | 116 | |
XM_017010485.1 | XP_016865974.1 | 11 - 12 | 2113 | 116 | |
XM_017010486.1 | XP_016865975.1 | 12 - 13 | 1992 | 116 | |
XM_017010487.1 | XP_016865976.1 | 10 - 11 | 1658 | 116 | |
XM_017010488.1 | XP_016865977.1 | 11 - 12 | 2519 | 116 | |
XM_017010489.1 | XP_016865978.1 | 12 - 13 | 2034 | 116 | |
XM_017010490.1 | XP_016865979.1 | 11 - 12 | 1904 | 116 | |
XM_017010491.1 | XP_016865980.1 | 11 - 12 | 1865 | 116 | |
XM_017010492.1 | XP_016865981.1 | 10 - 11 | 1909 | 116 | |
GenBank mRNA | AB023153.1 | - | 10 - 11 | 1605 | 116 |
AF152469.1 | - | 10 - 11 | 1686 | 116 | |
AF225919.1 | - | 10 - 11 | 1548 | 116 | |
AK074892.1 | - | 11 - 12 | 1799 | 116 | |
BC136420.1 | - | 9 - 10 | 1379 | 116 | |
BC136421.1 | - | 9 - 10 | 1379 | 116 | |
BC152464.1 | - | 10 - 11 | 1605 | 116 |