Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014920.3 | NP_055735.1 | 2 - 3 | 474 | 92 |
NM_016513.4 | NP_057597.2 | 3 - 4 | 585 | 92 | |
XM_011514419.1 | XP_011512721.1 | 2 - 3 | 474 | 92 | |
XM_011514420.2 | XP_011512722.1 | 3 - 4 | 616 | 92 | |
XM_011514421.1 | XP_011512723.1 | 3 - 4 | 555 | 92 | |
XM_017010485.1 | XP_016865974.1 | 3 - 4 | 848 | 92 | |
XM_017010486.1 | XP_016865975.1 | 4 - 5 | 727 | 92 | |
XM_017010487.1 | XP_016865976.1 | 2 - 3 | 393 | 92 | |
XM_017010488.1 | XP_016865977.1 | 3 - 4 | 1275 | 92 | |
XM_017010489.1 | XP_016865978.1 | 4 - 5 | 790 | 92 | |
XM_017010490.1 | XP_016865979.1 | 3 - 4 | 660 | 92 | |
XM_017010491.1 | XP_016865980.1 | 3 - 4 | 621 | 92 | |
XM_017010492.1 | XP_016865981.1 | 2 - 3 | 665 | 92 | |
GenBank mRNA | AB023153.1 | - | 2 - 3 | 361 | 92 |
AF152469.1 | - | 2 - 3 | 442 | 92 | |
AK074892.1 | - | 3 - 4 | 555 | 92 | |
BC035807.1 | - | 2 - 3 | 506 | 92 | |
BC136420.1 | - | 1 - 2 | 135 | 92 | |
BC136421.1 | - | 1 - 2 | 135 | 92 | |
BC152464.1 | - | 2 - 3 | 361 | 92 | |
DA456873.1 | - | 2 - 3 | 474 | 92 |