Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143935.1 | NP_001137407.1 | 16 - 17 | 1828 | 138 |
NM_021151.3 | NP_066974.2 | 15 - 16 | 1744 | 138 | |
XM_011516337.2 | XP_011514639.1 | 15 - 16 | 1744 | 138 | |
XM_017012370.1 | XP_016867859.1 | 16 - 17 | 1818 | 138 | |
GenBank mRNA | AF073770.1 | - | 15 - 16 | 1635 | 138 |
AF168793.1 | - | 15 - 16 | 1635 | 138 | |
AK127901.1 | - | 16 - 17 | 1784 | 138 | |
AK300114.1 | - | 10 - 11 | 1074 | 138 | |
AK313487.1 | - | 15 - 16 | 1687 | 138 | |
BC039004.1 | - | 15 - 16 | 1684 | 138 |