Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143935.1 | NP_001137407.1 | 8 - 9 | 983 | 102 |
NM_021151.3 | NP_066974.2 | 7 - 8 | 899 | 102 | |
XM_011516337.2 | XP_011514639.1 | 7 - 8 | 899 | 102 | |
XM_017012370.1 | XP_016867859.1 | 8 - 9 | 973 | 102 | |
GenBank mRNA | AF073770.1 | - | 7 - 8 | 790 | 102 |
AF168793.1 | - | 7 - 8 | 790 | 102 | |
AK127901.1 | - | 8 - 9 | 939 | 102 | |
AK300114.1 | - | 2 - 3 | 229 | 102 | |
BC039004.1 | - | 7 - 8 | 839 | 102 |