Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184732.1 | NP_001171661.1 | 10 - 11 | 1650 | 65 |
NM_001184733.1 | NP_001171662.1 | 10 - 11 | 1398 | 65 | |
NM_001184736.1 | NP_001171665.1 | 9 - 10 | 1313 | 65 | |
NM_004254.3 | NP_004245.2 | 10 - 11 | 1671 | 65 | |
XM_011545364.1 | XP_011543666.1 | 9 - 10 | 1246 | 65 | |
GenBank mRNA | AB042505.1 | - | 10 - 11 | 1653 | 65 |
AK298341.1 | - | 10 - 11 | 1368 | 65 | |
AK298351.1 | - | 11 - 12 | 1623 | 65 | |
AK307850.1 | - | 9 - 10 | 1278 | 65 | |
AK313187.1 | - | 10 - 11 | 1644 | 65 | |
BC022387.1 | - | 10 - 11 | 1665 | 65 |