Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033117.2 | NP_001028289.1 | 16 - 17 | 2580 | 65 |
NM_014850.3 | NP_055665.1 | 16 - 17 | 2652 | 65 | |
XM_011534295.1 | XP_011532597.1 | 17 - 18 | 2667 | 65 | |
XM_011534296.1 | XP_011532598.1 | 18 - 19 | 2784 | 65 | |
XM_011534301.2 | XP_011532603.1 | 14 - 15 | 4496 | 65 | |
XM_011534303.1 | XP_011532605.1 | 12 - 13 | 1392 | 65 | |
XM_017007574.1 | XP_016863063.1 | 16 - 17 | 2990 | 65 | |
XM_017007575.1 | XP_016863064.1 | 16 - 17 | 2918 | 65 | |
XM_017007576.1 | XP_016863065.1 | 18 - 19 | 2159 | 65 | |
XM_017007577.1 | XP_016863066.1 | 18 - 19 | 2098 | 65 | |
XM_017007578.1 | XP_016863067.1 | 16 - 17 | 3018 | 65 | |
XM_017007579.1 | XP_016863068.1 | 16 - 17 | 2986 | 65 | |
XM_017007580.1 | XP_016863069.1 | 18 - 19 | 2147 | 65 | |
GenBank mRNA | AB007871.2 | - | 16 - 17 | 1804 | 65 |
AF427144.1 | - | 16 - 17 | 2020 | 65 | |
AF464189.1 | - | 16 - 17 | 2108 | 65 | |
BC039300.1 | - | 19 - 20 | 2327 | 65 |