Assay Design



Gene Transcripts

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
Chromosome Location
Chr.14: 77274956 - 77320885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013382.5 NP_037514.2 10 - 11 1379 77
XM_011536675.2 XP_011534977.1 10 - 11 1379 77
XM_011536676.2 XP_011534978.1 9 - 10 1292 77
XM_011536677.2 XP_011534979.1 6 - 7 921 77
XM_011536679.2 XP_011534981.1 6 - 7 581 77
XM_017021244.1 XP_016876733.1 9 - 10 1213 77
XM_017021245.1 XP_016876734.1 6 - 7 667 77
GenBank mRNA AB209114.1 - 10 - 11 1201 77
AF105020.1 - 10 - 11 1382 77
AY090480.1 - 10 - 11 1382 77
BC031651.1 - 10 - 11 1269 77
BM127609.1 - 4 - 5 303 77
CR627447.1 - 10 - 11 949 77

View Full Product Details