Assay Design



Gene Transcripts

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
Chromosome Location
Chr.14: 77274956 - 77320885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013382.5 NP_037514.2 11 - 12 1459 82
XM_011536675.2 XP_011534977.1 11 - 12 1459 82
XM_011536676.2 XP_011534978.1 10 - 11 1372 82
XM_011536677.2 XP_011534979.1 7 - 8 1001 82
XM_011536679.2 XP_011534981.1 7 - 8 661 82
XM_017021244.1 XP_016876733.1 10 - 11 1293 82
XM_017021245.1 XP_016876734.1 7 - 8 747 82
GenBank mRNA AB209114.1 - 11 - 12 1281 82
AF105020.1 - 11 - 12 1462 82
AK023804.1 - 5 - 6 2050 82
AK025962.1 - 1 - 2 78 82
AY090480.1 - 11 - 12 1462 82
BC031651.1 - 11 - 12 1349 82
CR627447.1 - 11 - 12 1029 82

View Full Product Details