Assay Design



Gene Transcripts

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
Chromosome Location
Chr.14: 77274956 - 77320885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013382.5 NP_037514.2 12 - 13 1539 67
XM_011536675.2 XP_011534977.1 12 - 13 1539 67
XM_011536676.2 XP_011534978.1 11 - 12 1452 67
XM_011536677.2 XP_011534979.1 8 - 9 1081 67
XM_011536679.2 XP_011534981.1 8 - 9 741 67
XM_017021244.1 XP_016876733.1 11 - 12 1373 67
XM_017021245.1 XP_016876734.1 8 - 9 827 67
GenBank mRNA AB209114.1 - 12 - 13 1361 67
AK023804.1 - 6 - 7 2130 67
AK025962.1 - 2 - 3 158 67
BC031651.1 - 12 - 13 1429 67
CR627447.1 - 12 - 13 1109 67

View Full Product Details