Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013382.5 | NP_037514.2 | 14 - 15 | 1782 | 66 |
XM_011536675.2 | XP_011534977.1 | 14 - 15 | 1782 | 66 | |
XM_011536676.2 | XP_011534978.1 | 13 - 14 | 1695 | 66 | |
XM_011536677.2 | XP_011534979.1 | 10 - 11 | 1324 | 66 | |
XM_011536679.2 | XP_011534981.1 | 10 - 11 | 984 | 66 | |
XM_017021244.1 | XP_016876733.1 | 13 - 14 | 1616 | 66 | |
XM_017021245.1 | XP_016876734.1 | 10 - 11 | 1070 | 66 | |
GenBank mRNA | AB209114.1 | - | 14 - 15 | 1604 | 66 |
AF105020.1 | - | 14 - 15 | 1785 | 66 | |
AK023804.1 | - | 8 - 9 | 2373 | 66 | |
AK025962.1 | - | 4 - 5 | 401 | 66 | |
AY090480.1 | - | 14 - 15 | 1785 | 66 | |
BC031651.1 | - | 14 - 15 | 1672 | 66 | |
BE797202.1 | - | - | 188 | 66 | |
CR627447.1 | - | 14 - 15 | 1352 | 66 |