Assay Design



Gene Transcripts

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
Chromosome Location
Chr.14: 77274956 - 77320885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013382.5 NP_037514.2 4 - 5 752 107
XM_011536675.2 XP_011534977.1 4 - 5 752 107
XM_011536676.2 XP_011534978.1 3 - 4 665 107
XM_017021244.1 XP_016876733.1 4 - 5 746 107
GenBank mRNA AB209114.1 - 4 - 5 574 107
AF105020.1 - 4 - 5 755 107
AY090480.1 - 4 - 5 755 107
BC031651.1 - 4 - 5 642 107
BX248027.1 - 3 - 4 645 107
CR627447.1 - 4 - 5 322 107

View Full Product Details