Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206961.1 | NP_001193890.1 | 2 - 3 | 319 | 65 |
NM_001293225.1 | NP_001280154.1 | 3 - 4 | 516 | 65 | |
NM_015900.3 | NP_056984.1 | 3 - 4 | 521 | 65 | |
NR_120610.1 | 3 - 4 | 521 | 65 | ||
XM_017006572.1 | XP_016862061.1 | 4 - 5 | 553 | 65 | |
GenBank mRNA | AF035268.1 | - | 3 - 4 | 486 | 65 |
AF035269.1 | - | 3 - 4 | 486 | 65 | |
AK222705.1 | - | 3 - 4 | 484 | 65 | |
AK301880.1 | - | 2 - 3 | 292 | 65 | |
AK313519.1 | - | 3 - 4 | 484 | 65 | |
BC035347.1 | - | 3 - 4 | 473 | 65 | |
BX647516.1 | - | 3 - 4 | 516 | 65 | |
DB189568.1 | - | 3 - 4 | 522 | 65 | |
U37591.1 | - | 3 - 4 | 481 | 65 |