Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134419.1 | NP_001127891.1 | 9 - 10 | 1227 | 96 |
NM_001134420.1 | NP_001127892.1 | 9 - 10 | 1355 | 96 | |
NM_003503.3 | NP_003494.1 | 9 - 10 | 1261 | 96 | |
XM_005271241.2 | XP_005271298.1 | 9 - 10 | 1223 | 96 | |
XM_005271244.2 | XP_005271301.1 | 8 - 9 | 1165 | 96 | |
XM_017002425.1 | XP_016857914.1 | 8 - 9 | 1127 | 96 | |
XM_017002426.1 | XP_016857915.1 | 8 - 9 | 1110 | 96 | |
XM_017002427.1 | XP_016857916.1 | 8 - 9 | 1069 | 96 | |
GenBank mRNA | AB003698.1 | - | 9 - 10 | 1228 | 96 |
AB209337.1 | - | 6 - 7 | 3355 | 96 | |
AF005209.1 | - | 8 - 9 | 1096 | 96 | |
AF015592.1 | - | 9 - 10 | 1241 | 96 | |
AK298963.1 | - | 8 - 9 | 1173 | 96 | |
AK312726.1 | - | 9 - 10 | 1317 | 96 | |
AY312583.1 | - | 1 - 2 | 66 | 96 | |
BC110526.1 | - | 9 - 10 | 1184 | 96 | |
BC110527.1 | - | 9 - 10 | 1218 | 96 | |
BC111044.1 | - | 9 - 10 | 1241 | 96 |