Assay Design



Gene Transcripts

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
Chromosome Location
Chr.1: 28887091 - 29120046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166005.1 NP_001159477.1 10 - 11 1583 92
NM_001166006.1 NP_001159478.1 10 - 11 1656 92
NM_001166007.1 NP_001159479.1 10 - 11 1566 92
NM_004437.3 NP_004428.1 11 - 12 1646 92
NM_203342.2 NP_976217.1 11 - 12 1663 92
NM_203343.2 NP_976218.1 9 - 10 1478 92
XM_005245753.1 XP_005245810.1 10 - 11 1658 92
XM_005245757.1 XP_005245814.1 10 - 11 1658 92
XM_005245760.1 XP_005245817.1 10 - 11 1658 92
XM_005245761.1 XP_005245818.1 10 - 11 1658 92
XM_005245763.1 XP_005245820.1 10 - 11 1658 92
XM_005245764.1 XP_005245821.1 10 - 11 1658 92
XM_005245765.1 XP_005245822.1 10 - 11 1658 92
XM_005245768.1 XP_005245825.1 10 - 11 1658 92
XM_005245769.1 XP_005245826.1 10 - 11 1658 92
XM_005245770.1 XP_005245827.1 10 - 11 1658 92
XM_005245772.4 XP_005245829.1 10 - 11 1658 92
XM_005245773.4 XP_005245830.1 10 - 11 1658 92
XM_005245774.1 XP_005245831.1 10 - 11 1658 92
XM_006710434.1 XP_006710497.1 10 - 11 1658 92
XM_006710439.1 XP_006710502.1 10 - 11 1658 92
XM_011540956.1 XP_011539258.1 10 - 11 1658 92
XM_011540957.1 XP_011539259.1 10 - 11 1658 92
XM_011540958.1 XP_011539260.1 10 - 11 1658 92
XM_011540959.1 XP_011539261.1 10 - 11 1658 92
XM_011540960.1 XP_011539262.1 10 - 11 1658 92
XM_011540961.1 XP_011539263.1 10 - 11 1658 92
XM_011540962.1 XP_011539264.1 10 - 11 1658 92
XM_011540963.2 XP_011539265.1 10 - 11 1658 92
XM_011540964.1 XP_011539266.1 10 - 11 1554 92
XM_011540965.2 XP_011539267.1 10 - 11 1658 92
XM_017000581.1 XP_016856070.1 9 - 10 1553 92
XM_017000582.1 XP_016856071.1 9 - 10 1553 92
XM_017000583.1 XP_016856072.1 10 - 11 1658 92
XM_017000584.1 XP_016856073.1 10 - 11 1949 92
XM_017000585.1 XP_016856074.1 10 - 11 1946 92
XM_017000586.1 XP_016856075.1 10 - 11 1658 92
XM_017000587.1 XP_016856076.1 10 - 11 1658 92
XM_017000588.1 XP_016856077.1 10 - 11 1658 92
XM_017000589.1 XP_016856078.1 9 - 10 1553 92
XM_017000590.1 XP_016856079.1 9 - 10 1553 92
XM_017000591.1 XP_016856080.1 10 - 11 1658 92
XM_017000592.1 XP_016856081.1 10 - 11 1658 92
XM_017000593.1 XP_016856082.1 10 - 11 1554 92
XM_017000594.1 XP_016856083.1 10 - 11 1554 92
XM_017000595.1 XP_016856084.1 10 - 11 1554 92
XM_017000596.1 XP_016856085.1 10 - 11 1554 92
XM_017000597.1 XP_016856086.1 10 - 11 1554 92
XM_017000598.1 XP_016856087.1 10 - 11 1554 92
XM_017000599.1 XP_016856088.1 10 - 11 1554 92
XM_017000600.1 XP_016856089.1 10 - 11 1554 92
XM_017000601.1 XP_016856090.1 10 - 11 1554 92
XM_017000602.1 XP_016856091.1 10 - 11 1554 92
XM_017000603.1 XP_016856092.1 10 - 11 1554 92
XM_017000604.1 XP_016856093.1 10 - 11 1554 92
GenBank mRNA AB209649.1 - 10 - 11 1566 92
AF156225.1 - 9 - 10 1456 92
BC039079.1 - 10 - 11 1653 92
BC096104.1 - 7 - 8 872 92
BC096105.1 - 9 - 10 1190 92
BC113841.1 - 9 - 10 1190 92
BC114450.1 - 9 - 10 1190 92
J03796.1 - 9 - 10 1397 92
M14993.1 - 11 - 12 1627 92
M61733.1 - 11 - 12 1644 92

View Full Product Details