Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006499.4 | NP_006490.3 | 7 - 8 | 908 | 222 |
NM_201543.2 | NP_963837.1 | 7 - 8 | 908 | 96 | |
NM_201544.2 | NP_963838.1 | 6 - 7 | 965 | 96 | |
NM_201545.2 | NP_963839.1 | 7 - 8 | 778 | 222 | |
XM_011544188.2 | XP_011542490.1 | 6 - 7 | 722 | 222 | |
XM_017001274.1 | XP_016856763.1 | 6 - 7 | 722 | 96 | |
GenBank mRNA | AB209914.1 | - | 5 - 6 | 641 | 96 |
AF074000.1 | - | 7 - 8 | 907 | 96 | |
AF074001.1 | - | 7 - 8 | 907 | 222 | |
AF074002.1 | - | 7 - 8 | 907 | 225 | |
AF342815.1 | - | 5 - 6 | 553 | 96 | |
AF342816.1 | - | 5 - 6 | 553 | 222 | |
AF468213.1 | - | 5 - 6 | 527 | 222 | |
AK098613.1 | - | 6 - 7 | 721 | 222 | |
BC015818.1 | - | 6 - 7 | 676 | 222 | |
BC016486.2 | - | 6 - 7 | 915 | 96 | |
L78132.1 | - | 5 - 6 | 580 | 96 | |
X91790.1 | - | 5 - 6 | 569 | 96 |