Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000922.3 | NP_000913.2 | 4 - 5 | 1764 | 112 |
XM_006718249.3 | XP_006718312.1 | 4 - 5 | 1796 | 112 | |
XM_017017911.1 | XP_016873400.1 | 4 - 5 | 1796 | 112 | |
XM_017017912.1 | XP_016873401.1 | 4 - 5 | 1796 | 112 | |
GenBank mRNA | AY459346.1 | - | 4 - 5 | 1702 | 112 |
BC136565.1 | - | 4 - 5 | 1657 | 112 | |
BC136566.1 | - | 4 - 5 | 1657 | 112 | |
BC144248.1 | - | 4 - 5 | 1504 | 112 | |
BC150307.1 | - | 4 - 5 | 1764 | 112 | |
X95520.1 | - | 4 - 5 | 1411 | 112 |