Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289010.1 | NP_001275939.1 | 10 - 11 | 1978 | 98 |
NM_020379.3 | NP_065112.1 | 10 - 11 | 1978 | 98 | |
XM_017001860.1 | XP_016857349.1 | 9 - 10 | 1081 | 98 | |
GenBank mRNA | AB209275.1 | - | 10 - 11 | 1326 | 98 |
AF261655.1 | - | 10 - 11 | 1978 | 98 | |
BC137017.1 | - | 10 - 11 | 1784 | 98 |